Learn More
TA Instruments Gold Sample Pans Hermetic, DSC Gold Hermetic Pans, High Pressure Materials, up to 6 Atm, -180°C to 725°C, Use with Gold Hermetic Lids, 10 Pack
Catalog No. 5000580853 Shop All TA Instruments Products
Encompass_Preferred
Waters TA Instruments Gold Sample Pans, Hermetic are DSC gold pans designed for evaluating materials under higher self-generated pressures up to 6 atm (600 kPa gage). They have a temperature range of -180°C to 725°C and are used with gold hermetic lids (P/N: 900872.901).
Catalog No. 50-005-80853 Supplier TA Instruments Supplier No. 900871.901
Waters TA Instruments Gold Sample Pans, Hermetic are DSC gold pans designed for evaluating materials under higher self-generated pressures up to 6 atm (600 kPa gage). They have a temperature range of -180°C to 725°C and are used with gold hermetic lids (P/N: 900872.901).