Learn More
Electron Microscopy Sciences Gold Seal Microscope Slides Plain, 3 x 1", 1 mm thick
SDP
Catalog No. NC1390098
Encompass_Preferred
GOLDSEAL slides are made from pure white glass (sodalime) and at accurately cut sizes. To preserve the highest quality from manufacturer to ultimate user, all GOLD SEAL slides are hermetically sealed in our Moisture-Barrier system, providing a dry-slide environments during shipment and storage.
Size: 3 x 1"
Thickness: 1 mm
Catalog No. NC1390098 Supplier Electron Microscopy Sciences Supplier No. 6371005
GOLDSEAL slides are made from pure white glass (sodalime) and at accurately cut sizes. To preserve the highest quality from manufacturer to ultimate user, all GOLD SEAL slides are hermetically sealed in our Moisture-Barrier system, providing a dry-slide environments during shipment and storage.
Size: 3 x 1"
Thickness: 1 mm