Learn More
Electron Microscopy Sciences Gold Shadowed Latex 0.091 µm
SDP
Catalog No. 5026324
Encompass_Preferred
These latex particles have a diameter of 0.204um and are shadowed with a coating of gold. Measure the finest migrated gold particles in the shadow to determine the resolution of the STEM. Gold shadowed latex on formvar/carbon film, mounted on a copper grid, 3.05 mm diameter.
Catalog No. 50-263-24 Supplier Electron Microscopy Sciences Supplier No. 80040
These latex particles have a diameter of 0.204um and are shadowed with a coating of gold. Measure the finest migrated gold particles in the shadow to determine the resolution of the STEM. Gold shadowed latex on formvar/carbon film, mounted on a copper grid, 3.05 mm diameter.