Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gold standard solution, for AAS, 1 mg/ml Au in 10-20% HCl

Catalog No. p-220945
Change view
Click to view available options
Quantity:
100 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC196001000 100 mL
AC196005000 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC196001000 Supplier Thermo Scientific Chemicals Supplier No. 196001000

This product is not available for sale online. Please call Customer Service at 1-800-766-7000 or send an email to cs.webemail@thermofisher.com for a quote.

missing translation for 'validMainframeMsg'

CAS: 7440-57-5 | Au | 196.97 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

For atomic adsorption spectroscopy
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Gold standard solution
Name Note 1 mg/ml Au in 0.5N HCl
Packaging Plastic bottle
CAS 7440-57-5, 7647-01-0, 7732-18-5
Density 1.0900g/mL
Specific Gravity 1.09
Physical Form Powder and/or Chunks
Molecular Formula Au
Concentration 990 to 1010 μg/mL of Au(3+) in 10 to 20% HCl
MDL Number MFCD00003436
Color Orange/Yellow to Yellow
Solubility Soluble in water
InChI Key PCHJSUWPFVWCPO-UHFFFAOYSA-N
SMILES [Au]
IUPAC Name gold
Molecular Weight (g/mol) 196.97
Quantity 100 mL
PubChem CID 23985
ChEBI CHEBI:30050
Formula Weight 196.97
Grade AAS
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.