Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gold Wire, 0.025mm (0.001 in.) dia., 99.95% (Metals basis), Alfa Aesar™

Form: Approx.0.01g/m

Manufacturer:  Alfa Aesar 40585G8

Quantity 20m

 View more versions of this product

Catalog No. AA40585G8


  Check Availability
Add to cart

Specifications Description & Specifications

Chemical Identifiers

7440-57-5
99.95% (Metals basis)
Gold
Wire, 0.025mm (0.001 in.) dia.
MFCD00003436

Specifications

20m
Gold colored wire
99.95% (Metals basis)
Odorless
Gold
7440-57-5
Wire, 0.025mm (0.001 in.) dia.
MFCD00003436
Gold colored
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit