Gold Wire, 0.35mm (0.0138 in.) dia., Premion™, 99.999% (Metals basis), Alfa Aesar™

Form: Approx.1.82g/m

Manufacturer: Alfa Aesar 45084CC

Quantity 500cm

 View more versions of this product

Catalog No. AA45084CC

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 7440-57-5
    Physical Form Wire, 0.35mm (0.0138 in.) dia.
    Assay Percent Range 99.999% (Metals basis)
    MDL Number MFCD00003436
    Chemical Name or Material Gold

    Specifications

    Quantity 500cm
    Color Gold colored
    Physical Form Wire, 0.35mm (0.0138 in.) dia.
    Assay Percent Range 99.999% (Metals basis)
    MDL Number MFCD00003436
    CAS 7440-57-5
    Odor Odorless
    Appearance Gold colored wire
    Chemical Name or Material Gold

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit