Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gold wire, 14kt, red, 1.63mm (0.064 in.) dia., Au 58.3% min, Thermo Scientific™

Catalog No. p-7051037
Change view
Click to view available options
Quantity:
10 cm
25 cm
50 cm
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA40328BS 10 cm
AA40328BU 25 cm
AA40328BW 50 cm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AA40328BS Supplier Thermo Scientific Chemicals Supplier No. 40328BS
Only null left
Add to Cart

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Specifications

Color Red
Chemical Name or Material Gold wire (14kt)
Name Note 1.63mm (0.064 in.) dia.
Quantity 10 cm
Weight ≈28g/m
Form Wire
MDL Number MFCD00003436
Solubility Information Insoluble in water.
Concentration or Composition (by Analyte or Components) % Concentration Cu>Ag, Au ≤58.3%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.