Learn More
Sigma Aldrich Fine Chemicals Biosciences Gold wire, length 1 m, 0.20 mm diameter, high purity 99.99% | 7440-57-5 | MFCD00003436 | 1EA
Catalog No. 501851369
Encompass
Gold wire, length 1 m, 0.20 mm diameter, high purity 99.99% | Purity: 99.99% | Mol Wt: 196.97 | 7440-57-5 | MFCD00003436 | 1EA
Catalog No. 50-185-1369 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. GF783174001EA
Gold wire, length 1 m, 0.20 mm diameter, high purity 99.99% | Purity: 99.99% | Mol Wt: 196.97 | 7440-57-5 | MFCD00003436 | 1EA