Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gold Zinc slugs, 3.175mm (0.125in)dia x 3.175mm (0.125in) length, 99.95% (metals basis)

Catalog No. AA43470KJ
Change view
Click to view available options
Quantity:
1 Pc.
5 Pc.
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA43470KJ 5 Pc.
AA43470KG 1 Pc.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA43470KJ Supplier Thermo Scientific Chemicals Supplier No. 043470KJ
Add to Cart

missing translation for 'validMainframeMsg'

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Gold Zinc slugs
Name Note 3.175mm (0.125 in.) dia. x 3.175mm (0.125 in.) length
Avg. Mol. Wt. or Mol. Wt. Range Au:Zn; 90:10 wt%
Quantity 5 Pc.
Weight ≈0.45g/slug
Form Slug
Assay Percent Notes (metals basis)
MDL Number MFCD02092916
Percent Purity 99.95%

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.