Learn More
Roboz Surgical Roboz Golden A-5 Clipper, No. 10 Blade, 1 Ea
SDP
Catalog No. 5000574441
Encompass
The Golden A-5 clipper is a professional-grade grooming and surgical instrument featuring a detachable No. 10 blade designed for versatile performance. Engineered for reliable operation, this unit is double insulated and certified for safety in demanding environments.
  • Operates on 120 volts AC power
  • Includes a detachable No. 10 blade
  • Features double-insulated construction for safety
  • UL listed for compliance
  • Designed to meet OSHA specifications
  • Weighs 26 oz.
Catalog No. 50-005-74441 Supplier Roboz Surgical Supplier No. RC5905
The Golden A-5 clipper is a professional-grade grooming and surgical instrument featuring a detachable No. 10 blade designed for versatile performance. Engineered for reliable operation, this unit is double insulated and certified for safety in demanding environments.
  • Operates on 120 volts AC power
  • Includes a detachable No. 10 blade
  • Features double-insulated construction for safety
  • UL listed for compliance
  • Designed to meet OSHA specifications
  • Weighs 26 oz.