Learn More
New England Biolabs, Inc. NEB Golden Gate Assembly Kit (BsmBI-v2) – 20 reactions
SDP
Catalog No. NC1828212
Encompass
The NEB Golden Gate Assembly Kit (BsmBI-v2) contains an optimized mix of BsmBI-v2 and T4 DNA Ligase. BsmBI-v2 has been engineered by NEB and outperforms BsmBI in Golden Gate Assemblies. Together these enzymes can direct the assembly of multiple inserts/modules and also single insert/library generation cloning with single insert(s) using the Golden Gate approach. The pGGAselect destination plasmid provides a backbone for your assembly. It has flanking recognition sites in the correct orientation for BsmBI-directed assemblies, and also BsaI- and BbsI-directed assemblies, enabling the destination plasmid to conveniently be used with the most commonly used Type IIS restriction enzymes used for Golden Gate Assembly. Please note that while general descriptions regarding Golden Gate Assembly use the BsmBI nomenclature, this kit and protocols feature the specific engineered form optimized for Golden Gate Assembly, BsmBI-v2.
Catalog No. NC1828212 Supplier New England Biolabs, Inc. Supplier No. E1602S
The NEB Golden Gate Assembly Kit (BsmBI-v2) contains an optimized mix of BsmBI-v2 and T4 DNA Ligase. BsmBI-v2 has been engineered by NEB and outperforms BsmBI in Golden Gate Assemblies. Together these enzymes can direct the assembly of multiple inserts/modules and also single insert/library generation cloning with single insert(s) using the Golden Gate approach. The pGGAselect destination plasmid provides a backbone for your assembly. It has flanking recognition sites in the correct orientation for BsmBI-directed assemblies, and also BsaI- and BbsI-directed assemblies, enabling the destination plasmid to conveniently be used with the most commonly used Type IIS restriction enzymes used for Golden Gate Assembly. Please note that while general descriptions regarding Golden Gate Assembly use the BsmBI nomenclature, this kit and protocols feature the specific engineered form optimized for Golden Gate Assembly, BsmBI-v2.