Learn More
Electron Microscopy Sciences EMS Golden Nylon Brushes, set of 4
SDP
Catalog No. NC1637564
Encompass_Preferred
These brushes are perfect for cryomicrotomy – for controlled manipulation of frozen sections in the cryostat, as well as cryomicrotome. Easier to clean than animal hair brushes and more durable.
Catalog No. NC1637564 Supplier Electron Microscopy Sciences Supplier No. 6611005
These brushes are perfect for cryomicrotomy – for controlled manipulation of frozen sections in the cryostat, as well as cryomicrotome. Easier to clean than animal hair brushes and more durable.