Learn More
NANOPROBES INC GOLDENHANCE BLOTS 48ML TOTAL
Catalog No. NC1716263
Encompass
Gold developer specially formulated for use with protein blots. Comes in four solutions, to be mixed just before use. 48 mL total, enough for 6 (7 x 8.4 cm) Westerns.Initiator/Moderator/ Activator/Buffer:12 mL of each(48 mL total)
Catalog No. NC1716263 Supplier NANOPROBES INC Supplier No. 211548 ML
Gold developer specially formulated for use with protein blots. Comes in four solutions, to be mixed just before use. 48 mL total, enough for 6 (7 x 8.4 cm) Westerns.Initiator/Moderator/ Activator/Buffer:12 mL of each(48 mL total)