Learn More
NANOPROBES INC GOLDENHANCE LM GELM 40ML TOTAL
Catalog No. NC1716259
Encompass
Gold developer formulated for use with light microscopy samples. Comes in four solutions, to be mixed just before use. 28 mL total, enough for 250 slides (100 microliters/slide). Initiator/Moderator/ Activator/Buffer:7 mL of each(40 mL total).
Catalog No. NC1716259 Supplier NANOPROBES INC Supplier No. 211228 ML
Gold developer formulated for use with light microscopy samples. Comes in four solutions, to be mixed just before use. 28 mL total, enough for 250 slides (100 microliters/slide). Initiator/Moderator/ Activator/Buffer:7 mL of each(40 mL total).