Learn More
Medipoint GOLDENROD 7MM LANCET- 1000/CS
Catalog No. NC9089910
Encompass_Preferred
Goldenrod Animal Lancets are sterile, individually wrapped, avail in 7 different pt lengths to safely draw blood from the Temporal Facial Vein of Mice and Rats, the Ear Vein of Pigs and Rabbits, and the Brachial Vein of Chickens.
Catalog No. NC9089910 Supplier Medipoint Supplier No. GOLDENROD7MM
Goldenrod Animal Lancets are sterile, individually wrapped, avail in 7 different pt lengths to safely draw blood from the Temporal Facial Vein of Mice and Rats, the Ear Vein of Pigs and Rabbits, and the Brachial Vein of Chickens.