Learn More
Medipoint GOLDENROD LANCET 3MM 1000/CS
Catalog No. NC0024199
Encompass_Preferred
Goldenrod Animal Lancets are sterile, individually wrapped, avail in 7 different pt lengths to safely draw blood from the Temporal Facial Vein of Mice and Rats, the Ear Vein of Pigs and Rabbits, and the Brachial Vein of Chickens.
Catalog No. NC0024199 Supplier Medipoint Supplier No. GOLDENROD3MM
Goldenrod Animal Lancets are sterile, individually wrapped, avail in 7 different pt lengths to safely draw blood from the Temporal Facial Vein of Mice and Rats, the Ear Vein of Pigs and Rabbits, and the Brachial Vein of Chickens.