Learn More
World Precision Instrument Pins, 0.031" Gold-plated (pkg of 50)
SDP
Catalog No. NC0102069
Encompass
Gold-Plated Electrode Tips or electrode connectors play a crucial role in creating a reliable electrical connection between wires and the experimental apparatus, particularly in the field of neuroscience or other disciplines involving electrical measurements or stimulation. These are the tips of wires that come into direct contact with the experimental subject, such as neural tissue. The tips coated in gold, a conductive material, to ensure good electrical conductivity. Gold has excellent conductivity, is corrosion resistance, and is biocompatible, making it suitable for applications where precise and stable electrical connections are essential. These electrode tips connect amplifiers, recording devices, or stimulation equipment. In electrophysiological experiments, the gold-plated tips may be part of headstages. Headstages are components that connect to electrodes and are often mounted on the subject, enabling the acquisition of neural signals.
Catalog No. NC0102069 Supplier World Precision Instrument Supplier No. 5482
Gold-Plated Electrode Tips or electrode connectors play a crucial role in creating a reliable electrical connection between wires and the experimental apparatus, particularly in the field of neuroscience or other disciplines involving electrical measurements or stimulation. These are the tips of wires that come into direct contact with the experimental subject, such as neural tissue. The tips coated in gold, a conductive material, to ensure good electrical conductivity. Gold has excellent conductivity, is corrosion resistance, and is biocompatible, making it suitable for applications where precise and stable electrical connections are essential. These electrode tips connect amplifiers, recording devices, or stimulation equipment. In electrophysiological experiments, the gold-plated tips may be part of headstages. Headstages are components that connect to electrodes and are often mounted on the subject, enabling the acquisition of neural signals.