Learn More
Perkin Elmer US LLC Gold-Plated Copper Sealing Disks - Qty. 10
Catalog No. 502112446
Encompass_Preferred
When combined with the proper seals, the high-pressure capsules permit the study of explosive materials. They can be used in any situation where the advantage of a self-generating atmosphere is to be employed. They can withstand a maximum internal pressure of 150 atmospheres and have broad temperature range.
Catalog No. 50-211-2446 Supplier Perkin Elmer US LLC Supplier No. B0182905
When combined with the proper seals, the high-pressure capsules permit the study of explosive materials. They can be used in any situation where the advantage of a self-generating atmosphere is to be employed. They can withstand a maximum internal pressure of 150 atmospheres and have broad temperature range.