Learn More
Fine Science Tools Goldstein Retractor, Stainless Steel, 3 cm
SDP
Catalog No. NC9798314
Encompass
Product: Goldstein Retractor - 3x3 Sharp/3cm/1.8cm Maximum Spread/ Teeth: 3x3 Sharp/ Teeth Length: 3mm/ Teeth Width: 6.5mm/ Maximum Spread: 1.8cm/ Alloy: Stainless Steel/ Length: 3cm/ Unit: Each
Catalog No. NC9798314 Supplier Fine Science Tools Supplier No. 1700303
Product: Goldstein Retractor - 3x3 Sharp/3cm/1.8cm Maximum Spread/ Teeth: 3x3 Sharp/ Teeth Length: 3mm/ Teeth Width: 6.5mm/ Maximum Spread: 1.8cm/ Alloy: Stainless Steel/ Length: 3cm/ Unit: Each