Learn More
Aviva Systems Biology Corp GOLGA1 Antibody - N-terminal region (ARP54639_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP54639_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against GOLGA1. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the N terminal region of human GOLGA1. Alternate names: MGC33154, golgin-97ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-285-950