Learn More
Aviva Systems Biology Corp GOLGA5 Antibody - N-terminal region (ARP46573_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP46573_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against GOLGA5. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human GOLGA5. Alternate names: GOLIM5, RFG5, ret-IIApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-59350