Learn More
Aviva Systems Biology Corp GOLGA7 Antibody - N-terminal region (ARP56836_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP56836_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against GOLGA7. It was validated on Western Blot. GOLGA7,golgin A7,GCP16; GOLGA7A; HSPC041; MGC4876; MGC21096; GOLGA3AP1; GOLGA7,Golgin subfamily A member 7,Golgin subfamily A member 7,golgi autoantigen, golgin subfamily a, 7,Golgi complex-associated protein of 16kDa,golgi complex-associated protein of 16 kDaThe immunogen is a synthetic peptide directed towards the N terminal region of human GOLGA7. Alternate names: GCP16, GOLGA3AP1, HSPC041, MGC21096, MGC4876, GOLGA7AApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat, Zebrafish
Catalog No. 50-286-222