Learn More
Enzo Life Sciences GOLGI ID Green assay kit (1x1Kit)
SDP
Catalog No. NC0074031
Encompass_Preferred
Dependably  localizes to Golgi apparatus with minimal staining of ER. Applications: Fluorescence microscopy. Contents: GOLGI ID Green detection reagent: 50nmoles. Hoechst 33342 nuclear stain: 50μl. 10X assay buffer 1: 15ml. 50X assay buffer 2: 1.2ml
Catalog No. NC0074031 Supplier Enzo Life Sciences Supplier No. ENZ51028K100
Dependably  localizes to Golgi apparatus with minimal staining of ER. Applications: Fluorescence microscopy. Contents: GOLGI ID Green detection reagent: 50nmoles. Hoechst 33342 nuclear stain: 50μl. 10X assay buffer 1: 15ml. 50X assay buffer 2: 1.2ml