Learn More
enQuire Bioreagents Calponin-1 Anti-Human Monoclonal [Clone SPM169], Size: 100ug
Supplier:  enQuire Bioreagents MSM1X102P1
Encompass_Preferred
Product Type: Antibody. Application: Flow, IF, WB, ICC, IHC. Conjugate/Tag: Unconjugated. Clone: SPM581. Clonality: Monoclonal. Host: Mouse. Isotype: IgG1 kappa. Positive Control: Tonsil or lymph node. Cellular Staining: Golgi complex in cytoplasm. Buffer: 10mM PBS with 0.05% BSA & 0.05% azide. Concentration: 200ug/ml. Purity: Protein A/G Purified. Immunogen: SU-DHL-1 large cell lymphoma cells. Storage: Short term: Store at 2-4C. Long term, aliquot and store at -20C.
Catalog No. 50-188-5432