Learn More
Medchemexpress LLC HY-100540 10mg Medchemexpress, Golgicide A CAS:1139889-93-2 Purity:>98%
SDP
Catalog No. 501967744
Encompass_Preferred
Medchemexpress, HY-100540 10mg Golgicide A CAS:1139889-93-2 Golgicide A is a potent, highly specific, and reversible inhibitor of the cis-Golgi ADP-ribosylation factor guanine nucleotide exchange factors (ArfGEF), GBF1. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-7744 Supplier Medchemexpress LLC Supplier No. HY10054010MG
Medchemexpress, HY-100540 10mg Golgicide A CAS:1139889-93-2 Golgicide A is a potent, highly specific, and reversible inhibitor of the cis-Golgi ADP-ribosylation factor guanine nucleotide exchange factors (ArfGEF), GBF1. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.