Learn More
Medchemexpress LLC Golgicide A | 1139889-93-2 | MFCD27937048 | >98.0% | 284.30 g/mol | C17H14F2N2 | 50 MG
SDP
Catalog No. 5000214503
Encompass_Preferred
Golgicide A is a small-molecule, reversible inhibitor of the cis-Golgi ADP-ribosylation factor guanine nucleotide exchange factor GBF1, used as a research reagent to probe Golgi function and viral replication. It is intended for laboratory research use only.
  • Potent, highly specific, reversible inhibitor of GBF1.
  • Reduces replication of coxsackievirus B3 and other enteroviruses.
  • Useful for studying Golgi trafficking, membrane dynamics, and virus-host interactions.
  • Typical purity ≥98% suitable for biochemical and cell-based assays.
  • Molecular weight 284.30 g/mol; formula C17H14F2N2; CAS 1139889-93-2.
  • Supplied in formats appropriate for laboratory use; research use only.
Catalog No. 50-002-14503 Supplier Medchemexpress LLC Supplier No. HY10054050MG
Golgicide A is a small-molecule, reversible inhibitor of the cis-Golgi ADP-ribosylation factor guanine nucleotide exchange factor GBF1, used as a research reagent to probe Golgi function and viral replication. It is intended for laboratory research use only.
  • Potent, highly specific, reversible inhibitor of GBF1.
  • Reduces replication of coxsackievirus B3 and other enteroviruses.
  • Useful for studying Golgi trafficking, membrane dynamics, and virus-host interactions.
  • Typical purity ≥98% suitable for biochemical and cell-based assays.
  • Molecular weight 284.30 g/mol; formula C17H14F2N2; CAS 1139889-93-2.
  • Supplied in formats appropriate for laboratory use; research use only.