Learn More
Medchemexpress LLC Golgicide A-2 | 1394285-50-7 | 99.5% | 284.30 g/mol | C17H14F2N2 | 10 MG
SDP
Catalog No. 5000214211
Encompass_Preferred
Golgicide A-2 is a synthetic derivative of Golgicide A that selectively inhibits GBF1, an Arf guanine nucleotide exchange factor. Supplied as a research reagent, it is used to study Golgi function, vesicular trafficking, and antiviral mechanisms against enteroviruses.
  • Selective GBF1 (ArfGEF) inhibitor used to probe Golgi function.
  • Useful in vesicular trafficking and membrane dynamics studies.
  • Applied in antiviral research against enteroviruses and coxsackievirus.
  • High purity supports reproducible experimental results.
  • Available in small quantities suitable for laboratory assays.
Catalog No. 50-002-14211 Supplier Medchemexpress LLC Supplier No. HY100540B10MG
Golgicide A-2 is a synthetic derivative of Golgicide A that selectively inhibits GBF1, an Arf guanine nucleotide exchange factor. Supplied as a research reagent, it is used to study Golgi function, vesicular trafficking, and antiviral mechanisms against enteroviruses.
  • Selective GBF1 (ArfGEF) inhibitor used to probe Golgi function.
  • Useful in vesicular trafficking and membrane dynamics studies.
  • Applied in antiviral research against enteroviruses and coxsackievirus.
  • High purity supports reproducible experimental results.
  • Available in small quantities suitable for laboratory assays.