Learn More
Apexbio Technology LLC Golgicide A, 5mg. Cas: 1139889-93-2 MFCD: MFCD04128902. GBF1 inhibitor, potent, reversible and highly specific
SDP
Supplier:  Apexbio Technology LLC B13855
Encompass_Preferred
Golgicide A is a potent, reversible and highly specific GBF1 inhibitor.GBF1, the ArfGEF responsible for Arf1 activation and COPI recruitment to cis-Golgi MEMs, plays an important role in coordinating bidirectional transport and maintaining structural integrity of the Golgi [1]. Other sizes are available. Please inqury us for quote.
Catalog No. 50-190-5226