Learn More
Pyramex™ Goliath™ Safety Eyewear
Catalog No. 19156007 Shop All Pyramex Safety Products Products
Change view
Click to view available options
Frame Color:
Black
White
Lens Coating:
Anti-fog, Scratch-resistant, UV-treated
Scratch-resistant, UV-treated
Lens Tint:
Blue, Mirror
Clear
Gray
Indoor/Outdoor Mirror, Mirror
Orange, Mirror
Red, Mirror
Silver, Mirror
11 product options available for selection
Product selection table with 11 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Frame Color Lens Coating Lens Tint
19-156-007 White Scratch-resistant, UV-treated Blue, Mirror
19-155-753 Black Scratch-resistant, UV-treated Clear
19-155-754 Black Anti-fog, Scratch-resistant, UV-treated Clear
19-155-756 Black Anti-fog, Scratch-resistant, UV-treated Gray
19-155-757 Black Scratch-resistant, UV-treated Gray
19-155-758 Black Scratch-resistant, UV-treated Orange, Mirror
19-155-759 Black Scratch-resistant, UV-treated Blue, Mirror
19-155-760 Black Scratch-resistant, UV-treated Silver, Mirror
19-155-761 Black Scratch-resistant, UV-treated Indoor/Outdoor Mirror, Mirror
19-156-006 White Scratch-resistant, UV-treated Red, Mirror
19-156-005 White Scratch-resistant, UV-treated Gray
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 11 options available.
11 options
This item is not returnable. View return policy
Catalog No. 19-156-007 Supplier Pyramex™ Supplier No. SW5665D
Add to Cart
Add to Cart
This item is not returnable. View return policy
Goliath™ Safety Eyewear is suitable for both working environments and sports activities.
  • Comfort-fit, rubber temples prevent eyewear from slipping
  • Scratch resistant polycarbonate lens provides 99% UVA/B/C protection
Specifications
Certifications/Compliance ANSI Z87.1, CSA Z94.3-07
Lens Coating Scratch-resistant, UV-treated
Lens Type Polarized
Lens Tint Blue, Mirror
Product Type Full Frame Glasses
Frame Color White
Weight (Metric) 35 g
Material Polycarbonate
Disposable Reusable
Lens Material Polycarbonate
Frame Material Polycarbonate, TPR
UV Protection U6
Frame Type Full Frame
Autoclavable Autoclavable