Learn More
Aviva Systems Biology Corp GOLM1 Antibody - N-terminal region (ARP47269_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP47269_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against GOLM1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human GOLM1. Alternate names: C9orf155, FLJ22634, FLJ23608, GOLPH2, GP73, PSEC0257, bA379P1.3ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-59503