Learn More
ABclonal Technology GOLPH2 Rabbit PolymAb
SDP
Catalog No. 5000010770
Encompass_Preferred
The Golgi complex plays a key role in the sorting and modification of proteins exported from the endoplasmic reticulum The protein encoded by this gene is a type II Golgi transmembrane protein It processes proteins synthesized in the rough endoplasmic reticulum and assists in the transport of protein cargo through the Golgi apparatus The expression of this gene has been observed to be upregulated in response to viral infection Alternatively spliced transcript variants encoding the same protein have been described for this gene
Catalog No. 50-000-10770 Supplier ABclonal Technology Supplier No. A26409PM20UL
The Golgi complex plays a key role in the sorting and modification of proteins exported from the endoplasmic reticulum The protein encoded by this gene is a type II Golgi transmembrane protein It processes proteins synthesized in the rough endoplasmic reticulum and assists in the transport of protein cargo through the Golgi apparatus The expression of this gene has been observed to be upregulated in response to viral infection Alternatively spliced transcript variants encoding the same protein have been described for this gene