Learn More
Aviva Systems Biology Corp GOLPH3 Antibody - middle region (ARP62176_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP62176_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against GOLPH3. It was validated on Western Blot. . Alternate names: FLJ90675, GOPP1, GPP34, MIDASApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-287-599