Learn More
Selleck Chemical LLC Golvatinib (E7050) S2859-5mg
SDP
Catalog No. 503111672
Encompass_Preferred
Golvatinib (E7050) is a dual c-Met and VEGFR-2 inhibitor with IC50 of 14 nM and 16 nM does not inhibit bFGF-stimulated HUVEC growth (up to 1000 nM) Phase 1/2
Catalog No. 50-311-1672 Supplier Selleck Chemical LLC Supplier No. S28595MG
Golvatinib (E7050) is a dual c-Met and VEGFR-2 inhibitor with IC50 of 14 nM and 16 nM does not inhibit bFGF-stimulated HUVEC growth (up to 1000 nM) Phase 1/2