Learn More
Apexbio Technology LLC Golvatinib (E7050) 928037-13-2 10mM (in 1mL DMSO)
SDP
Catalog No. 5000062626
Encompass_Preferred
Golvatinib (E7050 CAS 928037-13-2) is a small-molecule inhibitor targeting the tyrosine kinases c-Met and VEGFR-2 Golvatinib inhibits phosphorylation of c-Met in MKN45 gastric cancer cells and VEGFR-2 in human umbilical vein endothelial cells (HUVECs) with IC50 values of 14 nM and 16 nM respectively Additionally it suppresses proliferation of various tumor cells including MKN45 EBC-1 Hs746T and SNU-5 and inhibits tumor growth in xenograft models via reduction of c-Met and VEGFR-2-mediated pathways Golvatinib is utilized in cancer research to explore targeted antitumor efficacy related to angiogenesis and oncogenic signaling
Catalog No. 50-000-62626 Supplier Apexbio Technology LLC Supplier No. B14395.1
Golvatinib (E7050 CAS 928037-13-2) is a small-molecule inhibitor targeting the tyrosine kinases c-Met and VEGFR-2 Golvatinib inhibits phosphorylation of c-Met in MKN45 gastric cancer cells and VEGFR-2 in human umbilical vein endothelial cells (HUVECs) with IC50 values of 14 nM and 16 nM respectively Additionally it suppresses proliferation of various tumor cells including MKN45 EBC-1 Hs746T and SNU-5 and inhibits tumor growth in xenograft models via reduction of c-Met and VEGFR-2-mediated pathways Golvatinib is utilized in cancer research to explore targeted antitumor efficacy related to angiogenesis and oncogenic signaling