Learn More
Med Vet International Golytely Powder Pack, Makes 4 Liters
Catalog No. 502828526
Encompass_Preferred
GoLYTELY is indicated for bowel cleansing prior to colonoscopy and barium enema X-ray examination. GoLYTELY is the original 4-liter GI lavage prep. GoLYTELY has proven safety in a wide range of patients, including diabetic, hypertensive, cardiac and renal patients as well as those with electrolyte imbalances.
Catalog No. 50-282-8526 Supplier Med Vet International Supplier No. RXGOLYTELY
GoLYTELY is indicated for bowel cleansing prior to colonoscopy and barium enema X-ray examination. GoLYTELY is the original 4-liter GI lavage prep. GoLYTELY has proven safety in a wide range of patients, including diabetic, hypertensive, cardiac and renal patients as well as those with electrolyte imbalances.