Learn More
TARGETMOL CHEMICALS INC Gomisin D 5MG
SDP
Catalog No. 502879324
Encompass_Preferred
Also available in 1 mL, 1 mg, 10 mg, 25 mg, 50 mg and bulk. Please contact Fisher for quotes. Gomisin D is a natural product.Gomisin D is used as a quality marker of Shengmai San and shenqi Jiangtang Granule.Gomisin D is a potential antidiabetic and anti-Alzheimer’s agent. Purity 98.2%
Catalog No. 50-287-9324 Supplier TARGETMOL CHEMICALS INC Supplier No. TN10365MG
Also available in 1 mL, 1 mg, 10 mg, 25 mg, 50 mg and bulk. Please contact Fisher for quotes. Gomisin D is a natural product.Gomisin D is used as a quality marker of Shengmai San and shenqi Jiangtang Granule.Gomisin D is a potential antidiabetic and anti-Alzheimer’s agent. Purity 98.2%