Learn More
Medchemexpress LLC Gomisin E | 72960-21-5 | 1 MG
SDP
Supplier:  Medchemexpress LLC HYN73101MG
Encompass_Preferred
Gomisin E is a dibenzocyclooctadiene lignan isolated from the fruits of *Schizandra chinensis*. It inhibits NFAT transcription with an IC50 of 4.73 μM.
  • Isolated from the fruits of *Schizandra chinensis*.
  • Inhibits NFAT transcription.
Catalog No. 50-004-17620