Learn More
Medchemexpress LLC HY-N0385 1mg Medchemexpress, Gomisin J CAS:66280-25-9 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYN03851MG
Encompass_Preferred
Medchemexpress, HY-N0385 1mg Gomisin J CAS:66280-25-9 Gomisin J is a small molecular weight lignan found in Schisandra chinensis and has been demonstrated to have vasodilatory activity. Gomisin J suppresses lipid accumulation by regulating the expression of lipogenic and lipolytic enzymes and inflammatory molecules through activation of AMPK, LKB1 and Ca2+/calmodulin-dependent protein kinase II and inhibition of fetuin-A in HepG2 cells. gomisin J has potential benefits in treating nonalcoholic fatty liver disease. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-197-0264