Learn More
Apexbio Technology LLC Gonadorelin Acetate 71447-49-9 10mg
SDP
Catalog No. 5000088467
Encompass_Preferred
Gonadorelin Acetate is a synthetic peptide agonist targeting gonadotropin-releasing hormone (GnRH) receptors It is designed to bind specifically to GnRH receptors on pituitary gonadotrophs thereby modulating secretion of luteinizing hormone (LH) and follicle-stimulating hormone (FSH) Gonadorelin Acetate exerts its biological activity primarily through receptor-mediated regulation of the reproductive hormone axis Based on these pharmacological properties Gonadorelin Acetate holds research potential in studies of reproductive hormone signaling pathways endocrine regulation associated with infertility disorders and characterization of gonadotropin responses in cellular or animal models
Catalog No. 50-000-88467 Supplier Apexbio Technology LLC Supplier No. C644210
Gonadorelin Acetate is a synthetic peptide agonist targeting gonadotropin-releasing hormone (GnRH) receptors It is designed to bind specifically to GnRH receptors on pituitary gonadotrophs thereby modulating secretion of luteinizing hormone (LH) and follicle-stimulating hormone (FSH) Gonadorelin Acetate exerts its biological activity primarily through receptor-mediated regulation of the reproductive hormone axis Based on these pharmacological properties Gonadorelin Acetate holds research potential in studies of reproductive hormone signaling pathways endocrine regulation associated with infertility disorders and characterization of gonadotropin responses in cellular or animal models