Learn More
Cayman Chemical Gonadorelin (acetate), Purity: ≥98%, CAS: 34973-08-5, 10 mg
SDP
Catalog No. 502936417
Encompass_Preferred
A tropic hormone and agonist of GNRHR (Ki = 13 nM in CHO cells expressing the human receptor); stimulates LH and FSH release from rat anterior pituitary cultures (0.1 μg); stimulates LH release from bovine anterior pituitary cultures (1 μM); induces ovulation in rats pretreated with fluphenazine (ED50 = 610.3 ng/kg)
Catalog No. 50-293-6417 Supplier Cayman Chemical Supplier No. 2424010 MG
A tropic hormone and agonist of GNRHR (Ki = 13 nM in CHO cells expressing the human receptor); stimulates LH and FSH release from rat anterior pituitary cultures (0.1 μg); stimulates LH release from bovine anterior pituitary cultures (1 μM); induces ovulation in rats pretreated with fluphenazine (ED50 = 610.3 ng/kg)