Learn More
Mitegen Goniometer Head Key, For Precise Crystal Alignment & Adjustment.
SDP
Catalog No. 501941113
Encompass
This Goniometer Head Key is an essential precision tool designed to facilitate accurate adjustments of goniometer heads in crystallography workflows. It is indispensable for researchers and technicians performing crystal mounting and alignment procedures for X-ray diffraction analysis. The specialized design of this key ensures a secure fit and reliable torque, allowing for precise rotational and translational movements of the goniometer head. This meticulous control is crucial for optimally orienting delicate crystals in the X-ray beam, directly impacting the quality and resolution of diffraction data. By enabling fine adjustments, this tool minimizes the risk of misalignment and contributes to enhanced experimental reproducibility. Its robust construction ensures durability and consistent performance in demanding laboratory environments, making it a vital accessory for achieving high-quality structural determination.
Catalog No. 501941113 Supplier Mitegen Supplier No. MR1013159
This Goniometer Head Key is an essential precision tool designed to facilitate accurate adjustments of goniometer heads in crystallography workflows. It is indispensable for researchers and technicians performing crystal mounting and alignment procedures for X-ray diffraction analysis. The specialized design of this key ensures a secure fit and reliable torque, allowing for precise rotational and translational movements of the goniometer head. This meticulous control is crucial for optimally orienting delicate crystals in the X-ray beam, directly impacting the quality and resolution of diffraction data. By enabling fine adjustments, this tool minimizes the risk of misalignment and contributes to enhanced experimental reproducibility. Its robust construction ensures durability and consistent performance in demanding laboratory environments, making it a vital accessory for achieving high-quality structural determination.