Learn More
Med Vet International Goniometer Plastic 360, J0752P
Catalog No. 5000583957
Encompass_Preferred
The goniometer can be used for the calculation of tibal plateau angles, wedge resections, angles of anteversion plus others. It also can be used to measure pre- and post-op range of motion to evaluate post-op physiotherapy. White background assures ease of seeing number on any color dog.
Catalog No. 50-005-83957 Supplier Med Vet International Supplier No. JOR752P
The goniometer can be used for the calculation of tibal plateau angles, wedge resections, angles of anteversion plus others. It also can be used to measure pre- and post-op range of motion to evaluate post-op physiotherapy. White background assures ease of seeing number on any color dog.