Learn More
Sigma Aldrich Gonzales High Speed PLA 3D Printing Filament
Catalog No. 503125715 Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
1 Ea.
Catalog No. Quantity
50-312-5715 1 Ea.
1 options
Catalog No. 50-312-5715 Supplier Sigma Aldrich Supplier No. TRD3D00211EA
Only null left
Add to Cart
Add to Cart
MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.
ParametersExtruder (nozzle) temperature: 200 +/-10°CBed temperature: cold to 80°CPrint speed: up to 200 mm/sFan: 20-100 %Minimum nozzle diameter: 0.35 mmPlate materials: glass with glue, blue tape, kapton tape, buildtak
Specifications
Quantity 1 Ea.