Learn More
Electron Microscopy Sciences Good Pastures Stain Solution 500 ML
SDP
Supplier:  Electron Microscopy Sciences 26031
Encompass_Preferred
Goodpasture’s Stain (MacCallum) solution is a prepared, ready-to-use, high quality staining solution that saves time and effort and produces consistent uniform results.
Catalog No. 50-319-59