Learn More
Innovative Research Inc Goose RBCs Packed 100 15ml
Catalog No. 502744522
Encompass_Preferred
Goose Red Blood Cells Packed 100% 15ml - Goose Red Blood Cells Packed 100% from Innovative Research are manufactured from whole blood collections from animals located in the US. The whole blood is washed to remove the platelet-rich plasma, buffy coat layer, and leukocytes. Red blood cells are perishable and are collected and processed upon receipt of order. This product is supplied as a 100% solution.
Catalog No. 50-274-4522 Supplier Innovative Research Inc Supplier No. IGGSRBC100P15ML
Goose Red Blood Cells Packed 100% 15ml - Goose Red Blood Cells Packed 100% from Innovative Research are manufactured from whole blood collections from animals located in the US. The whole blood is washed to remove the platelet-rich plasma, buffy coat layer, and leukocytes. Red blood cells are perishable and are collected and processed upon receipt of order. This product is supplied as a 100% solution.