Learn More
Dino Lite RK-02 Desk Clamp Flex Arm Mount
Catalog No. NC1641078
Encompass_Preferred
Dino-Lite stand RK-02 offers excellent shaping capability of the gooseneck arm. RK-02 has increased stability in articulation compared to other gooseneck stands. RK-02 allows a Dino-Lite microscope to easily position by bending the arm and by adjusting the rotating head. The arm base can also be rotated for additional viewing angles, or be fixed with the quick release fastener for uneven surface usage. The C-clamp of RK-02 fits to most desk surfaces or poles up to 3-inch in thickness, horizontally or vertically, with a 23 inch (60 cm) reach.
Catalog No. NC1641078 Supplier Dino Lite Supplier No. RK02
Dino-Lite stand RK-02 offers excellent shaping capability of the gooseneck arm. RK-02 has increased stability in articulation compared to other gooseneck stands. RK-02 allows a Dino-Lite microscope to easily position by bending the arm and by adjusting the rotating head. The arm base can also be rotated for additional viewing angles, or be fixed with the quick release fastener for uneven surface usage. The C-clamp of RK-02 fits to most desk surfaces or poles up to 3-inch in thickness, horizontally or vertically, with a 23 inch (60 cm) reach.