Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ GoPure™ MabCaptureC™ Screening Plate, 96-well, 20 μL
GREENER_CHOICE

Catalog No. A65812 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
1 Plate
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A65812 1 Plate
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A65812 Supplier Thermo Scientific™ Supplier No. A65812
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Thermo Scientific™ GoPure™ MabCaptureC™ 96-well screening plates contain MabCaptureC Affinity Matrix and provide a versatile platform for sample preparation, handling, and analysis.

GoPure MabCaptureC™ 96-well screening plates contain MabCaptureC Affinity Matrix and provide a versatile platform for sample preparation, handling, and analysis. They accommodate multiple samples simultaneously to achieve high throughput and data generation.

These pre-filled screening plates have the following features:

  • High-throughput screening capability enabling multiple conditions to be evaluated in a single plate experiment
  • 20 μL per well, helping preserve sample while maximizing data generation
  • Ready-to-use 96-well format for efficient evaluation of Thermo Scientific resins

Specifications

Concentration 220 μL Resin Slurry/Well
Content And Storage Store at 2°C to 8°C. Do not freeze.
Shipping Condition Wet Ice
Particle Size 70 μm
Form Resin Slurry
Product Line MabCaptureC
Quantity 1 Plate
For Use With (Application) Chromatography
Type Affinity Screening Plate
Sample Volume 20 μL
Ligand Type Engineered rProtein A
Format 96-well Filter Plate
Capacity (Metric) 800 μL
Show More Show Less

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.