Learn More
Medchemexpress LLC Goralatide acetate | 120081-14-3 | 99.1% | 50 MG
SDP
Catalog No. 5000340654
Encompass_Preferred
Goralatide is a natural tetrapeptide that functions as a physiological regulator of hematopoiesis. It has the ability to inhibit the proliferation of primitive haematopoietic cells. This product is available as a solid with a white to off-white appearance, a molecular weight of 547.56, and a formula of C22H37N5O11. It is intended for research use only and is not for human use.
  • Physiological regulator of hematopoiesis.
  • Inhibits primitive haematopoietic cell proliferation.
  • Available as a white to off-white solid.
  • High purity of 99.12%.
Catalog No. 50-003-40654 Supplier Medchemexpress LLC Supplier No. HY129283B50MG
Goralatide is a natural tetrapeptide that functions as a physiological regulator of hematopoiesis. It has the ability to inhibit the proliferation of primitive haematopoietic cells. This product is available as a solid with a white to off-white appearance, a molecular weight of 547.56, and a formula of C22H37N5O11. It is intended for research use only and is not for human use.
  • Physiological regulator of hematopoiesis.
  • Inhibits primitive haematopoietic cell proliferation.
  • Available as a white to off-white solid.
  • High purity of 99.12%.