Learn More
Cayman Chemical Goralatide (acetate), Purity: ≥95%, 10 mg
SDP
Catalog No. 502911829
Encompass_Preferred
A tetrapeptide regulator of hematopoiesis; decreases the proportion of mouse HPP-CFCs in S phase when cultured using regenerating femoral marrow conditioned medium at 1 ng/ml; reduces 5-FU-induced decreases in the number of HPP-CFCs, BFU-E cells, and CFU-GMs in mice at 4 µg/kg twice per day; reduces doxorubicin-induced mortality in mice
Catalog No. 50-291-1829 Supplier Cayman Chemical Supplier No. 3620710 MG
A tetrapeptide regulator of hematopoiesis; decreases the proportion of mouse HPP-CFCs in S phase when cultured using regenerating femoral marrow conditioned medium at 1 ng/ml; reduces 5-FU-induced decreases in the number of HPP-CFCs, BFU-E cells, and CFU-GMs in mice at 4 µg/kg twice per day; reduces doxorubicin-induced mortality in mice