Learn More
Aviva Systems Biology Corp GORASP1 Antibody - N-terminal region (ARP57679_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP57679_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against GORASP1. It was validated on Western Blot. GORASP1,golgi reassembly stacking protein 1, 65kDa,P65; GOLPH5; GRASP65; FLJ23443; MGC118894; MGC118897; GORASP1,Golgi reassembly-stacking protein 1,Golgi reassembly-stacking protein 1,OTTHUMP00000208961,Golgi phosphoprotein 5,Golgi peripheral membrane protein p65,Golgi reassembly and stacking protein 1,golgi reassembly-stacking protein of 65 kDa,Golgi reassembly and stacking protein, 65 kDaThe immunogen is a synthetic peptide directed towards the N terminal region of human GORASP1. Alternate names: FLJ23443, GOLPH5, GRASP65, MGC118894, MGC118897, P65ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-286-348